missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfate, 99+%

Product Code. 10050870 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
5 L
Packaging:
Plastic bottle
Unit Size:
1 L
100mL
5 L
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 10050870

Brand: Thermo Scientific Acros 116820010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 77-78-1 | C2H6O4S | 126.126 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-78-1
MDL Number MFCD00008416
InChI Key VAYGXNSJCAHWJZ-UHFFFAOYSA-N
Synonym dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfate
PubChem CID 6497
ChEBI CHEBI:59050
IUPAC Name dimethyl sulfate
SMILES COS(=O)(=O)OC

Specifications

Color Colorless
Density 1.3300g/mL
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C2H6O4S
Linear Formula (CH3)2SO4
Refractive Index 1.3860 to 1.3880 (20°C, 589nm)
Quantity 1 L
Fieser 01,293; 04,188; 13,239
Molecular Weight (g/mol) 126.126
Formula Weight 126.13
Percent Purity ≥99.0% (hydrolysis)
Physical Form Liquid
Chemical Name or Material Dimethyl sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.