missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfate, 99+%, Thermo Scientific Chemicals

Product Code. 10667953
Click to view available options
Quantity:
100 mL
1 L
5 L
Packaging:
Plastic bottle
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Alkylation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-78-1
MDL Number MFCD00008416
InChI Key VAYGXNSJCAHWJZ-UHFFFAOYSA-N
Synonym dimethyl sulphate, sulfuric acid, dimethyl ester, dimethyl monosulfate, dimethylsulfate, dimethylsulfat, sulfate dimethylique, sulfuric acid dimethyl ester, dimethylsulfaat, dimetilsolfato, dms methyl sulfate
PubChem CID 6497
ChEBI CHEBI:59050
IUPAC Name dimethyl sulfate
SMILES COS(=O)(=O)OC

Specifications

Melting Point -32.0°C
Color Colorless
Density 1.3300g/mL
Boiling Point 188.0°C
Flash Point 83°C
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Molecular Formula C2H6O4S
Linear Formula (CH3)2SO4
Refractive Index 1.3860 to 1.3880
Quantity 5 L
Beilstein 01,283
Fieser 01,293; 04,188; 13,239
Merck Index 15,328
Specific Gravity 1.33
Solubility Information Solubility in water: 2.8g/100mL (18°C). Other solubilities: hydrolysis rapidly at 18°C or above in water,soluble in ehter,dioxane,acetone and aromatic,hydrocarbons,sparingly soluble in carbon disulfide and,aliphatic hydrocarbons
Molecular Weight (g/mol) 126.13
Viscosity 1.81 mPa.s (20°C)
Formula Weight 126.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material Dimethyl sulfate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H314-Causes severe skin burns and eye damage.
  • H350-May cause cancer.
  • H330-Fatal if inhaled.
  • H341-Suspected of causing genetic defects.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'