missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals Product Code.: 11398217

Dimethyl sulfide, 99+%, Thermo Scientific Chemicals

Product Code. 11398217
250 mL
Click to view available options
Quantity:
250 mL
1 L
This item is not returnable. View return policy

Product Code. 11398217

Brand: Thermo Scientific Alfa Aesar 022949.AK

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethyl sulfide is used as a displaceable ligand in chloro(dimethyl sulfide)gold(I) and other coordination compounds, as a reducing agent in ozonolysis reactions as well as a food flavoring component. Further, it is utilized for the preparation of dimethyl sulfoxide, which is a common and important industrial solvent. It finds application in the petroleum industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-18-3
Molecular Formula C2H6S
Molecular Weight (g/mol) 62.13
MDL Number MFCD00008562
InChI Key QMMFVYPAHWMCMS-UHFFFAOYSA-N
Synonym dimethyl sulfide, methyl sulfide, methane, thiobis, dimethylsulfide, dimethylsulphide, methyl thioether, 2-thiapropane, dimethyl sulphide, dimethyl thioether, methylthiomethaneShow MoreShow Less
PubChem CID 1068
ChEBI CHEBI:17437
SMILES CSC

Specifications

Melting Point -83°C
Density 0.845 g/mL
Boiling Point 38°C
Flash Point -48°C (-54°F)
Odor Unpleasant
Refractive Index 1.4351
Quantity 250 mL
UN Number UN1164
Merck Index 14,6123
Solubility Information Miscible with alcohols,ethers,esters,ketones,aliphatic and aromatic hydrocarbons. Slightly miscible with
IUPAC Name (methylsulfanyl)methane
Formula Weight 62.14
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material Dimethyl sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS27J073 Certificate TypeCertificate of Analysis Date04/07/2023 Catalog Number022949.K2,022949.AK,022949
Lot NumberU17I023 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog Number022949.K2,022949.AK
Lot NumberZ07H004 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number022949.K2,022949.AK
Lot Number10157710 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number022949.K2,022949.AK
Lot NumberD25Q31 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number022949.K2,022949.AK
compliance-icons compliance-icons
Product Identifier
  • Dimethyl sulfide
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Dimethyl sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl sulfide, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.