missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, For UV-spectroscopy, ≥99.8% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass Bottle
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 18.5°c |
Density | 1.10g/mL |
Boiling Point | 189°C |
Quantity | 250 mL |
Explosion Limit | 42% (63°F) |
Flash Point | 87°C (188.6°F) |
Ignition Point | 573°F |
Packaging | Glass Bottle |
Refractive Index | 1.479 |
Linear Formula | (CH3)2SO |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'