missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfOxide, ≥99.9% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Description
Honeywell Chromasolv™ GC-headspace solvents:
- developed particularly for the identification and quantification of residual solvents in pharmaceuticals using GC-headspace methods described in monographs from the United States Pharmacopeia (USP) or the European Pharmacopoeia (Ph. Eur.) and following the International Conference on Harmonization of Technical Requirements for Registration of Pharmaceuticals for Human Use (ICH) guidelines
- very pure solvents with extremely low concentrations of the defined residual solvents
- state-of-the art production and application tests
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.10g/mL |
Quantity | 2.5 L |
Explosion Limit | 42%, 63°F |
Flash Point | 87°C (188.6°F) |
Ignition Point | 573°F |
Linear Formula | (CH3)2SO |
Vapor Pressure | 0.42 mmHg (20°C) |
UN Number | NA1993 |
Beilstein | 506008 |
IUPAC Name | methylsulfinylmethane |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'