missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, ≥99.9%, Honeywell Riedel-de Haën™

Product Code. 15645630
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

ACS Reagent,≥99.9%

ACS GENERAL USE SOLVENTS

  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Color Colorless
Density 1.10g/mL
Quantity 100 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
IUPAC Name methanesulfinylmethane
Formula Weight 78.13
Percent Purity ≥99.9%
Grade ACS Reagent
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'