missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, ≥99.9%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
6 x 500 mL
4 x 4 L
6 x 1 L
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Color | Colorless |
Density | 1.10g/mL |
Quantity | 6 x 1 L |
Explosion Limit | 42%, 63°F |
Flash Point | 87°C (188.6°F) |
Ignition Point | 573°F |
Linear Formula | (CH3)2SO |
Vapor Pressure | 0.42 mmHg (20°C) |
Beilstein | 506008 |
IUPAC Name | methylsulfinylmethane |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'