missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, ≥99% (Molecular Biology/ACS Certified), MP Biomedicals
Click to view available options
Quantity:
100 mL
1 L
Specifications
Quantity | 1 L |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13 |
Grade | Certified ACS Reagent |
Chemical Name or Material | Dimethyl Sulfoxide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'