missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.1 L
Description
- Dimethyl sulfoxide is used to enhance dermal absorption of many chemicals.
- A solvent for many organic and inorganic compounds including fats, carbohydrates, dyes, resins, and polymers.
- Used in antifreeze or hydraulic fluids and as a cryopreservative for cell cultures.
- Used in the oxidation of thiols and disulfides to sulfonic acids.
- Used as a PCR cosolvent to help improve yields, especially in long PCR.

Specifications
Specifications
CAS | 67-68-5 |
Quantity | 0.1 L |
Molecular Formula | C2H6OS |
MDL Number | MFCD00002089 |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
SMILES | CS(C)=O |
IUPAC Name | methylsulfinylmethane |
Molecular Weight (g/mol) | 78.13 |
PubChem CID | 679 |
Show More |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Dimethyl Sulfoxide (99+% Purity), MP Biomedicals™ >
Spot an opportunity for improvement?Share a Content Correction