missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl Sulfoxide, Certified AR for Analysis, Fisher Chemical™
80.38 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
HDPE Bottle
Plastic Container
Description
C2H6OS, CAS Number-67-68-5, demasorb, demsodrox, methyl sulfoxide, dimexide, dimethyl sulfoxide, dimethylsulfoxide, demavet, methane, sulfinylbis, dimethyl sulphoxide, dmso, 2.5L, 189 deg.C, CHEBI:28262, Colorless, 78.13g/mol, IAZDPXIOMUYVGZ-UHFFFAOYSA-N, methylsulfinylmethane, 18.4 deg.C, 2089
Specifications
Melting Point | 18.4°C |
Color | Colorless |
Boiling Point | 189°C |
Quantity | 500 mL |
Packaging | HDPE Bottle |
Physical Form | Liquid |
Vapor Pressure | 0.55mbar at 20°C |
Viscosity | 1.98 mPaS at 25°C |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'