missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Description
- For HPLC, spectrophotometry, and environmental analysis
- Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
- Low UV impurity profile
- Low impurity gradient LC baseline
- Very low levels of non-volatile impurities
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 16°C to 19°C |
Density | 1.10g/mL |
Boiling Point | 189°C |
Quantity | 6 x 1 L |
Refractive Index | n20/D 1.479 |
Linear Formula | (CH3)2SO |
Vapor Pressure | 0.42mmHg (20°C) |
Beilstein | 506008 |
IUPAC Name | methylsulfinylmethane |
Formula Weight | 78.13g/mol |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'