Learn More
Dimethyl Sulfoxide, Fisher BioReagents™
Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %
£32.70 - £249.63
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methylsulfinylmethane |
SMILES | CS(C)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10103483
|
Fisher BioReagents
BP231-100 |
100 mL | Glass Bottle |
£32.70
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10499683
|
Fisher BioReagents
BP231-1 |
1 L | HDPE Bottle |
£150.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11463883
|
Fisher BioReagents
BP231-4 |
4 L | HDPE Bottle | N/A | ||||||
Description
Dimethyl Sulfoxide, >-99.7%, C2H6OS, CAS Number-67-68-5, methane, sulfinylbis, dmso, demasorb, dimethyl sulphoxide, demavet, dimethyl sulfoxide, demsodrox, dimethylsulfoxide, dimexide, methyl sulfoxide, 100mL, <-0.30 Absorbance Units at 280nm, Vapor Pressure: 0.55mbar at 20 deg.C, >-99.7 %
DMSO is a cryopreservation agent for cells, and may also be used in bacterial transformations.Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methylsulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(C)=O |
Specifications
18.4°C | |
67-68-5 | |
>190°C | |
189°C | |
Liquid | |
0.001% max. | |
(CH3)2SO | |
0.55mbar at 20°C | |
0.2% max. | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99.7% |
≤0.30 Absorbance Units at 280nm | |
Colorless | |
1.100g/cm³ | |
Pass Test | |
≥99.7 % | |
C2H6OS | |
MFCD00002089 | |
1.98 mPa/s at 25°C | |
15, 3285 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.13 | |
Dimethyl Sulfoxide |
Safety and Handling
WARNING!
Emergency Overview
Combustible liquid. DMSO readily penetrates skin and may carry other dissolved chemicals into the body. Hygroscopic. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.
NFPA
Health:2
Flammability:2
Instability:0
Recommended Storage : RT
- Methyl sulfoxide
Your input is important to us. Please complete this form to provide feedback related to the content on this product.