missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, Honeywell Riedel-de Haen™
Click to view available options
Quantity:
6 x 1 L
Packaging:
Glass Bottle
Description
Specifications
CAS | 67-68-5 |
Quantity | 6 x 1 L |
Packaging | Glass Bottle |
Physical Form | Liquid |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
Percent Purity | min. 99.9 % |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'