missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl Sulfoxide, Multi-Compendial, J.T. Baker™

Catalog Number 15538504
378.10 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
100 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Bottle
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
Quantity 100 mL
Flash Point 89.00°C
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.10kg
IUPAC Name methylsulfinylmethane
Formula Weight 78.13
Grade Multi-Compendial
Chemical Name or Material Dimethyl Sulfoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'