missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Dimethyl sulfoxide, puriss. p.a., Honeywell
Puriss. p.a., dried, ≤0.02% water
£127.04 - £229.28
Chemical Identifiers
CAS | 67-68-5 |
---|---|
Molecular Formula | C2H6OS |
Molecular Weight (g/mol) | 78.13 |
MDL Number | MFCD00002089 |
InChI Key | IAZDPXIOMUYVGZ-UHFFFAOYSA-N |
Synonym | dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide |
PubChem CID | 679 |
ChEBI | CHEBI:28262 |
IUPAC Name | methylsulfinylmethane |
SMILES | CS(C)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15644710
|
Honeywell Riedel-de Haën
34943-1L |
1 L |
£127.04
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654710
|
Honeywell Riedel-de Haën
34943-2.5L |
2.5 L |
£229.28
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
- It is the leading brand offering High-Purity Solvents for dedicated applications.
Chemical Identifiers
67-68-5 | |
78.13 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
679 | |
methylsulfinylmethane |
C2H6OS | |
MFCD00002089 | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CHEBI:28262 | |
CS(C)=O |
Specifications
67-68-5 | |
42%, 63°F | |
573°F | |
(CH3)2SO | |
0.42 mmHg (20°C) | |
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide | |
CS(C)=O | |
78.13 | |
CHEBI:28262 | |
≥99.9% (GC) | |
Dimethyl sulfoxide |
1.10g/mL | |
87°C (188.6°F) | |
C2H6OS | |
MFCD00002089 | |
506008 | |
IAZDPXIOMUYVGZ-UHFFFAOYSA-N | |
methylsulfinylmethane | |
679 | |
78.13g/mol | |
puriss. p.a. |
Safety and Handling
missing translation for 'einecsNumber' : 200-664-3
Product Identifier
- Dimethyl sulfoxide