missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Reagent Grade, Honeywell

Catalog Number 15687820
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
1 gA
4 L
5 L
25 L
230 kg
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
This item is not returnable. View return policy
67-68-5
C2H6OS
78.13
MFCD00002089
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
679
CHEBI:28262
CS(C)=O
Density 1.10g/mL
Quantity 1 gA
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
Beilstein 506008
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Grade Reagent
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Identifier
  • Dimethyl sulfoxide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'