missing translation for 'onlineSavingsMsg'
Learn More
Dimethyl sulfoxide, Reagent Grade, Honeywell Product Code.: 15667820

Dimethyl sulfoxide, Reagent Grade, Honeywell

Product Code. 15667820 Shop All Honeywell Research Chemicals Products
100 mL
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
1 gA
4 L
5 L
25 L
230 kg
This item is not returnable. View return policy

Product Code. 15667820

Brand: Honeywell Research Chemicals D5879100ML

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O

Specifications

Density 1.10g/mL
Quantity 100 mL
Explosion Limit 42%, 63°F
Flash Point 87°C (188.6°F)
Ignition Point 573°F
Linear Formula (CH3)2SO
Vapor Pressure 0.42 mmHg (20°C)
Beilstein 506008
IUPAC Name methylsulfinylmethane
Formula Weight 78.13g/mol
Percent Purity ≥99.5%
Grade Reagent
Chemical Name or Material Dimethyl sulfoxide
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyl sulfoxide, Reagent Grade, Honeywell >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.