missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulfoxide, Reagent Grade, Honeywell

£25.40 - £13505.80

Chemical Identifiers

CAS 67-68-5
Molecular Formula C2H6OS
Molecular Weight (g/mol) 78.13
MDL Number MFCD00002089
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
Synonym dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
PubChem CID 679
ChEBI CHEBI:28262
SMILES CS(C)=O
View More Specs

Products 9
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15667820
View Documents
Honeywell Research Chemicals
D5879-100ML
100 mL
£25.40
100mL
Please to purchase this item. Need a web account? Register with us today!
15637830
View Documents
Honeywell Research Chemicals
D5879-500ML
500 mL
£61.67
500mL
Please to purchase this item. Need a web account? Register with us today!
15697820
View Documents
Honeywell Research Chemicals
D5879-1L
1 L
£100.11
1 L
Please to purchase this item. Need a web account? Register with us today!
15607830
View Documents Promotion Details
Honeywell Research Chemicals
D5879-2.5L
2.5 L
£237.59
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15698480
View Documents
Honeywell Research Chemicals
D5879-4L
4 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15647830
View Documents
Honeywell Research Chemicals
D5879-5L
5 L
£478.41
5 L
Please to purchase this item. Need a web account? Register with us today!
15627830
View Documents
Honeywell Research Chemicals
D5879-25L
25 L
£1700.44
25 L
Please to purchase this item. Need a web account? Register with us today!
15687820
View Documents
Honeywell Research Chemicals
1 gA N/A
Please to purchase this item. Need a web account? Register with us today!
15617830
View Documents
Honeywell Research Chemicals
D5879-230KG
230 kg
£13505.80
230kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

67-68-5
78.13
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
679
CS(C)=O
C2H6OS
MFCD00002089
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CHEBI:28262

Specifications

67-68-5
42%, 63°F
573°F
(CH3)2SO
0.42 mmHg (20°C)
dimethyl sulfoxide, dmso, methyl sulfoxide, dimethylsulfoxide, dimethyl sulphoxide, methane, sulfinylbis, demsodrox, demasorb, demavet, dimexide
CS(C)=O
78.13
CHEBI:28262
≥99.5%
Dimethyl sulfoxide
1.10g/mL
87°C (188.6°F)
C2H6OS
MFCD00002089
506008
IAZDPXIOMUYVGZ-UHFFFAOYSA-N
methylsulfinylmethane
679
78.13g/mol
Reagent
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 200-664-3

SDS
Documents

Documents

Product Certifications