missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl sulphoxide, BAKER ANALYZED™, J.T. Baker™

Catalog Number 10198150
Click to view available options
1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Laboratory Reagent

Melting Point 18.5°C
CAS 67-68-5
Color Max. 10 (APHA)
Density 1.101 g/cm³ (20°C)
Boiling Point 189°C (1013 hPa)
Quantity 1 L
Flash Point 87°C
Packaging Glass Bottle
Assay Percent Range min. 99.0%
Residue after Evaporation max. 0.01%
Molecular Formula C2H6OS
MDL Number MFCD00002089
Water max. 0.2%
InChI Key IAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILES CS(C)=O
IUPAC Name methanesulfinylmethane
Molecular Weight (g/mol) 78.13
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'