missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl terephthalate-2,3,5,6-d4, 98 atom % D, Thermo Scientific Chemicals

Product Code. 15457987
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74079-01-9
Molecular Formula C10H10O4
Molecular Weight (g/mol) 198.21
MDL Number MFCD00182544
InChI Key WOZVHXUHUFLZGK-LNFUJOGGSA-N
Synonym dimethyl terephthalate-2,3,5,6-d4, 1,4-benzenedicarboxylic acid-d4 dimethyl ester, dimethyl p-phthalate-d4, dmt-d4, dimethyl terephthalate-d4, dimethyl p-benzenedicarboxylate-d4, dimethyl terephthalate-d4 ring-d4, methyl 4-carbomethoxy benzoate-d4, terephthalic acid-d4 dimethyl ester, dimethyl 1,4-benzenedicarboxylate-d4
PubChem CID 12241382
IUPAC Name dimethyl 2,3,5,6-tetradeuteriobenzene-1,4-dicarboxylate
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Quantity 1 g
Formula Weight 198.21
Percent Purity 95%
Assay 98 atom % D
Chemical Name or Material Dimethyl terephthalate-2,3,5,6-d4

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'