missing translation for 'onlineSavingsMsg'
Learn More

Dimethyl terephthalate, 99%, Thermo Scientific Chemicals

Product Code. 10012420
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 120-61-6
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.19
MDL Number MFCD00008440
InChI Key WOZVHXUHUFLZGK-UHFFFAOYSA-N
Synonym dimethyl terephthalate, dimethyl p-phthalate, 1,4-benzenedicarboxylic acid, dimethyl ester, dimethyl p-benzenedicarboxylate, di-me terephthalate, dimethyl 4-phthalate, terephthalic acid, dimethyl ester, dimethyl 1,4-benzenedicarboxylate, terephthalic acid dimethyl ester, methyl p-methoxycarbonyl benzoate
PubChem CID 8441
SMILES COC(=O)C1=CC=C(C=C1)C(=O)OC

Specifications

Acidity 0.03mg KOH/g max.
Melting Point 140°C to 143°C
Color White
Boiling Point 288°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 1 kg
Beilstein 09, 843
Merck Index 15, 9305
Solubility Information Solubility in water: 0.036g/L (20°C). Other solubilities: soluble in ether and hot alcohol
Sublimation Point 285.0 °C
IUPAC Name 1,4-dimethyl benzene-1,4-dicarboxylate
Formula Weight 194.19
Percent Purity 99%
Physical Form Flakes or Pellets
Chemical Name or Material Dimethyl terephthalate, 99%
Show More Show Less
Product Identifier
  • Dimethyl terephthalate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'