missing translation for 'onlineSavingsMsg'
Learn More

Dimethylaluminium chloride, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals

£76.38 - £531.65

Chemical Identifiers

CAS 1184-58-3, 142-82-5
MDL Number MFCD00000458
InChI Key JGHYBJVUQGTEEB-UHFFFAOYSA-M
Synonym aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution
PubChem CID 79147
IUPAC Name chloro(dimethyl)alumane
SMILES C[Al](C)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10290413
View Documents
Thermo Scientific Acros
377791000
100 mL AcroSeal™ Glass bottle
£76.38
100mL
Please to purchase this item. Need a web account? Register with us today!
10200423
View Documents Promotion Details
Thermo Scientific Acros
377798000
800 mL AcroSeal™ Glass Bottle
£531.65
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
Specifications

Chemical Identifiers

1184-58-3, 142-82-5
JGHYBJVUQGTEEB-UHFFFAOYSA-M
79147
C[Al](C)Cl
MFCD00000458
aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution
chloro(dimethyl)alumane

Specifications

1184-58-3, 142-82-5
0.7110g/mL
C2H6AlCl
15, 3249
aluminum, chlorodimethyl, dimethylaluminum chloride, dimethylaluminium chloride, chlorodimethylaluminum, dimethylaluminum chloride solution, 1.0 m in hexanes, chloro dimethyl alumane, dimethylaluminum chloride solution
JGHYBJVUQGTEEB-UHFFFAOYSA-M
chloro(dimethyl)alumane
79147
Solution
Colorless to Gray
−4°C
MFCD00000458
0.711
Solubility in water: vigorous reaction. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
C[Al](C)Cl
92.5
92.5
Dimethylaluminium chloride, 0.9M Solution in Heptane
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause drowsiness or dizziness.
Toxic to aquatic life with long lasting effects.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
In contact with water releases flamm

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
Do NOT induce vomiting.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 214-668-8

Product Identifier
  • Dimethylaluminium chloride, 0.9M solution in heptane

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1

Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
  • H336-May cause drowsiness or dizziness.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P331-Do NOT induce vomiting.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only