Learn More
Dimethylamine, 2M solution in methyl alcohol, AcroSeal™, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 370268000
View more versions of this product
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Dimethylamine, 2M solution in methanol, Acros Organics™ | |
AcroSeal™ Glass bottle | |
800 mL | |
ROSDSFDQCJNGOL-UHFFFAOYSA-N | |
N-methylmethanamine | |
674 | |
45.08 |
124-40-3, 67-56-1 | |
C2H7N | |
dimethylamine, n,n-dimethylamine, methanamine, n-methyl, dimethylamin, dimethyl amine, dimethyl-amine, dimethylamine anhydrous, rcra waste number u092, dimethylamine solution | |
CNC | |
45.085 | |
CHEBI:17170 |
Safety and Handling
- Dimethylamine, 2M solution in methyl alcohol
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H370-Causes damage to organs.
- H314-Causes severe skin burns and eye damage.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only