missing translation for 'onlineSavingsMsg'
Learn More

Dimethylamine, 40 wt.% solution in water, Thermo Scientific Chemicals

Product Code. 10647554 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10647554

Brand: Thermo Scientific Acros 163670100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Dimethylamine
CAS 124-40-3, 7732-18-5
Melting Point -37.0°C
Color Colorless
Density 0.8900 g/mL
Boiling Point 54.0°C
Flash Point -18°C
Packaging Plastic drum
Molecular Formula C2H7N
Linear Formula (CH3)2NH
Refractive Index 1.37
MDL Number MFCD00008288
Quantity 10 L
Beilstein 04, 39
Fieser 07,119
Merck Index 15, 3250
Specific Gravity 0.89
Synonym dimethylamine, n,n-dimethylamine, dimethylamine anhydrous, dimethylamin, dimethyl amine, rcra waste number u092, dimethyl-amine, methanamine, n-methyl, dimethylamine solution
Solubility Information Solubility aq. soln.: >500 g/L (20°C). Other solubilities: soluble in alcohol and ether
InChI Key ROSDSFDQCJNGOL-UHFFFAOYSA-N
SMILES CNC
IUPAC Name dimethylamine
Molecular Weight (g/mol) 45.09
PubChem CID 674
ChEBI CHEBI:17170
Formula Weight 45.07
Percent Purity ≥40%
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethylamine, solution in water
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Dimethylamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.