Learn More
Dimethyldioctadecylammonium bromide, 99+%, Thermo Scientific Chemicals
£33.50 - £351.00
Chemical Identifiers
CAS | 3700-67-2 |
---|---|
Molecular Formula | C38H80BrN |
Molecular Weight (g/mol) | 630.96 |
MDL Number | MFCD00041975 |
InChI Key | PSLWZOIUBRXAQW-UHFFFAOYSA-M |
Synonym | dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 |
PubChem CID | 77293 |
IUPAC Name | dimethyl(dioctadecyl)azanium;bromide |
SMILES | CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10447022
|
Thermo Scientific Acros
408260050 |
5 g | Glass bottle |
£33.50
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10147460
|
Thermo Scientific Acros
408260250 |
25 g | Glass bottle |
£111.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10005173
|
Thermo Scientific Acros
408261000 |
100 g | Glass bottle |
£351.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
3700-67-2 | |
630.96 | |
PSLWZOIUBRXAQW-UHFFFAOYSA-M | |
77293 | |
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] |
C38H80BrN | |
MFCD00041975 | |
dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 | |
dimethyl(dioctadecyl)azanium;bromide |
Specifications
3700-67-2 | |
White | |
Authentic | |
C38H80BrN | |
MFCD00041975 | |
1.047 | |
Solubility in water: .. Other solubilities: soluble in acetonitrile,methanol,ethanol,aceton,e,ethyl acetate | |
CCCCCCCCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCCCCCCCC.[Br-] | |
630.96 | |
630.96 | |
Granular Powder |
151.0°C to 157.0°C | |
1.0470g/mL | |
99.0 to 101% (Argentometry) | |
(CH3)2N(Br)((CH2)17CH3)2 | |
04, IV, 829 | |
dimethyldioctadecylammonium bromide, n,n-dimethyl-n-octadecyloctadecan-1-aminium bromide, dimethyldistearylammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide, unii-z5t47r065a, dsab, distearyldimethylammonium bromide, dimethyldioctadecylazanium bromide, distearyl dimethyl ammonium bromide, 1-octadecanaminium, n,n-dimethyl-n-octadecyl-, bromide 1:1 | |
PSLWZOIUBRXAQW-UHFFFAOYSA-M | |
dimethyl(dioctadecyl)azanium;bromide | |
77293 | |
99+% | |
Dimethyldioctadecylammonium bromide |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 223-037-6
- Dimethyldioctadecylammonium bromide
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only