missing translation for 'onlineSavingsMsg'
Learn More
Dimethyldiphenyltin, Thermo Scientific Chemicals Product Code.: 11384557

Dimethyldiphenyltin, Thermo Scientific Chemicals

Product Code. 11384557
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11384557

Brand: Thermo Scientific Alfa Aesar 039617.14

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1080-43-9
Molecular Formula C14H16Sn
Molecular Weight (g/mol) 302.992
MDL Number MFCD00058812
InChI Key INCQQSKGFIBXAY-UHFFFAOYSA-N
Synonym dimethyldiphenyltin, acmc-1bqk3, stannane, dimethyldiphenyl
PubChem CID 11130460
IUPAC Name dimethyl(diphenyl)stannane
SMILES C[Sn](C)(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Quantity 25 g
UN Number UN2788
Solubility Information Insoluble in water.
Formula Weight 302.97
Physical Form Liquid
Chemical Name or Material Dimethyldiphenyltin

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP07D023 Certificate TypeCertificate of Analysis Date24/06/2024 Catalog Number039617.06,039617.03,039617.14
Lot NumberX19C032 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039617.06,039617.03,039617.14
Lot NumberE03Z076 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039617.06,039617.03,039617.14
Lot NumberE04S015 Certificate TypeCertificate of Analysis Date24/11/2021 Catalog Number039617.06,039617.03,039617.14
Lot NumberD29U005 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog Number039617.06,039617.03,039617.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyldiphenyltin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.