missing translation for 'onlineSavingsMsg'
Learn More
Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Thermo Scientific™ Product Code.: 10039291

Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Thermo Scientific™

Product Code. 10039291
1 g, Amber Glass Bottle
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10039291

Brand: Thermo Scientific Maybridge SEW02190DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 73257-47-3
Molecular Formula C18H21NO5
Molecular Weight (g/mol) 331.368
MDL Number MFCD00085027
InChI Key IAXDEFZXLVTHLU-UHFFFAOYSA-N
Synonym dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 3,5-dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydro-3,5-pyridinedicarboxylate, maybridge1_007478, cbmicro_033161, 1,4-dihydro-2,6-dimethyl-4-4-methoxyphenyl-3,5-pyridinedicarboxylic acid dimethyl ester, dimethyl 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethylpyridine-3,5-dicarboxylate, dimethyl 4-4-methoxyphenyl-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxy late, 3,5-pyridinedicarboxylic acid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, dimethyl ester, 3,5-pyridinedicarboxylicacid, 1,4-dihydro-4-4-methoxyphenyl-2,6-dimethyl-, 3,5-dimethyl ester
PubChem CID 614332
IUPAC Name dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
SMILES CC1=C(C(C(=C(N1)C)C(=O)OC)C2=CC=C(C=C2)OC)C(=O)OC

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 331.37
Percent Purity 97%
Chemical Name or Material Dimethyl 4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyle4-(4-methoxyphenyl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.