missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DimethylFormamide, Certified AR for Analysis, Fisher Chemical™
141.13 GBP valid until 2024-12-31
Use promo code "23271" to get your promotional price.
Use promo code "23271" to get your promotional price.
Click to view available options
Quantity:
500 mL
1 L
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
HDPE plastic bottle
Plastic Coated Glass bottle
Plastic container
Description
DimethylFormamide, Certified AR for Analysis
Specifications
Melting Point | -61°C |
Color | Colorless |
Boiling Point | 153°C |
Packaging | Amber glass bottle |
Quantity | 2.5 L |
Formula Weight | 73.09g/mol |
Vapor Pressure | 4.9mbar at 20°C |
Physical Form | Liquid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'