missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DimethylFormamide, for HPLC, Fisher Chemical™
£97.10 - £208.00
Chemical Identifiers
CAS | 68-12-2 |
---|---|
Molecular Formula | C3H7NO |
Molecular Weight (g/mol) | 73.10 |
MDL Number | MFCD00003284 |
InChI Key | ZMXDDKWLCZADIW-UHFFFAOYSA-N |
Synonym | dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa |
PubChem CID | 6228 |
ChEBI | CHEBI:17741 |
IUPAC Name | N,N-dimethylformamide |
SMILES | CN(C)C=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10346180
|
Fisher Chemical
D/3846/15 |
1 L | Amber glass bottle |
£97.10
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10356180
|
Fisher Chemical
D/3846/17 |
2.5 L | Amber glass bottle |
£200.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10161660
|
Fisher Chemical
D/3846/PB17 |
2.5 L | Plastic Coated Glass bottle |
£208.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
DimethylFormamide, for HPLC
Chemical Identifiers
68-12-2 | |
73.10 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
6228 | |
N,N-dimethylformamide |
C3H7NO | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CHEBI:17741 | |
CN(C)C=O |
Specifications
68-12-2 | |
Colorless | |
C3H7NO | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
4.9mbar at 20°C |
-61°C | |
153°C | |
MFCD00003284 | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09g/mol | |
Liquid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Dimethylformamide
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.