Learn More
DimethylFormamide, for Peptide Synthesis, Packed Under Nitrogen, Fisher Chemical
Brand: Fisher Chemical D/3848/PB17
199.70 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 22
Additional Details : CAS Number : 68-12-2 Weight : 2.36250kg Transport : UN number : 2265 Chem class : 3 Pack group : III
Description
2.5L, Plastic Coated Glass bottle
Specifications
68-12-2 | |
Colorless | |
Plastic Coated Glass bottle | |
MFCD00003284 | |
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa | |
CN(C)C=O | |
73.10 | |
CHEBI:17741 | |
4.9mbar at 20°C |
-61°C | |
153°C | |
C3H7NO | |
2.5 L | |
ZMXDDKWLCZADIW-UHFFFAOYSA-N | |
N,N-dimethylformamide | |
6228 | |
73.09g/mol | |
Liquid |
Safety and Handling
- Dimethylformamide
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
Hazard Statement
- H226-Flammable liquid and vapour.
- H312-Harmful in contact with skin.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
- H360D-May damage the unborn child.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
Supplemental information
- MIXTURE LIST-Contains : dimethylformamide