Learn More
Dimethylglyoxime, 99+%, ACS reagent, Thermo Scientific Chemicals
£21.81 - £596.97
Chemical Identifiers
CAS | 95-45-4 |
---|---|
Molecular Formula | C4H8N2O2 |
Molecular Weight (g/mol) | 116.12 |
MDL Number | MFCD00002117 |
InChI Key | JGUQDUKBUKFFRO-GGWOSOGESA-N |
Synonym | biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky |
PubChem CID | 5356010 |
SMILES | C\C(=N/O)\C(\C)=N\O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10615491
|
Thermo Scientific Acros
408330250 |
25 g | Plastic bottle |
£21.81
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10625491
|
Thermo Scientific Acros
408331000 |
100 g | Plastic bottle |
£53.27
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10015700
|
Thermo Scientific Acros
408335000 |
500 g | Plastic bottle |
£159.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10751544
|
Thermo Scientific Acros
408330025 |
2.5 kg | Plastic bottle |
£596.97
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
95-45-4 | |
116.12 | |
JGUQDUKBUKFFRO-GGWOSOGESA-N | |
5356010 |
C4H8N2O2 | |
MFCD00002117 | |
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
C\C(=N/O)\C(\C)=N\O |
Specifications
95-45-4 | |
White | |
Authentic | |
C4H8N2O2 | |
MFCD00002117 | |
15, 3270 | |
Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene | |
JGUQDUKBUKFFRO-GGWOSOGESA-N | |
N-[(2Z)-3-nitrosobut-2-en-2-yl]hydroxylamine | |
5356010 | |
99+% | |
0.02% max. (in ethanol) | |
Dimethylglyoxime |
239.0°C to 242.0°C | |
0.05% max. | |
99+% | |
CH3C(=NOH)C(=NOH)CH3 | |
01, 772 | |
biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1ky | |
>70°C | |
C\C(=N/O)\C(\C)=N\O | |
116.12 | |
116.12 | |
ACS Reagent | |
Crystalline Powder |
Safety and Handling
GHS H Statement
Flammable solid.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediate
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 202-420-1
missing translation for 'rtecsNumber' : EK2975000
missing translation for 'tsca' : TSCA
- Dimethylglyoxime
Signal Word
- Danger
Hazard Category
- Flammable solid Category 2
- Acute toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
RUO – Research Use Only