missing translation for 'onlineSavingsMsg'
Learn More
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals Product Code.: 10408150

Dimethylglyoxime, 99+%, Thermo Scientific Chemicals

Product Code. 10408150
100 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10408150

Brand: Thermo Scientific Acros 172001000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethylglyoxime, 99+%, C4H8N2O2, CAS Number-95-45-4, 2,3-butanedione dioxime, chugaev's reagent, biacetyl, dioxime, 2,3-diisonitrosobutane, diacetyldioxime, nsc 9, diacetyl dioxime, unii-2971mft1ky, biacetyl dioxime, dimethylglyoxime, 500g, 01,772, White, 202-420-1, 116.12, GHS Signal Word: Danger

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-45-4
Molecular Formula C4H8N2O2
Molecular Weight (g/mol) 116.12
MDL Number MFCD00002117
InChI Key JGUQDUKBUKFFRO-GGWOSOGESA-N
Synonym biacetyl dioxime, dimethylglyoxime, diacetyldioxime, 2,3-butanedione dioxime, diacetyl dioxime, 2,3-diisonitrosobutane, biacetyl, dioxime, chugaev's reagent, nsc 9, unii-2971mft1kyShow MoreShow Less
PubChem CID 5356010
SMILES C\C(=N/O)\C(\C)=N\O

Specifications

Melting Point 239.0°C to 242.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99+%
Packaging Plastic bottle
Linear Formula CH3C(=NOH)C(=NOH)CH3
Quantity 100 g
Beilstein 01,772
Merck Index 15,327
Solubility Information Solubility in water: 0,6g/L 20°C. Other solubilities: soluble in alcohol,ether,pyridine and acetone,slightly soluble in benzene
Sublimation Point >70 °C
IUPAC Name N-[(2Z)-3-nitrosobut-2-en-2-yl]hydroxylamine
Formula Weight 116.12
Percent Purity 99+%
Insoluble Matter 0.05% max. (in alcohol)
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, 99+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0468945 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number172005000,172000025,172001000
Lot NumberA0463725 Certificate TypeCertificate of Analysis Date20/06/2024 Catalog Number172005000,172000025,172001000
Lot NumberA0459450 Certificate TypeCertificate of Analysis Date19/01/2024 Catalog Number172005000,172000025,172001000
Lot NumberA0403336 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number172005000,172000025,172001000
Lot NumberA0393424 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number172005000,172000025,172001000
compliance-icons compliance-icons
Product Identifier
  • Dimethylglyoxime
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P302+P352-IF ON SKIN: Wash with plenty of water/and soap.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethylglyoxime, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.