missing translation for 'onlineSavingsMsg'
Learn More

Dimethylglyoxime, disodium salt octahydrate, 99%

Product Code. 10146990 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
Packaging:
Glass bottle
Unit Size:
100g
1kg
250g
This item is not returnable. View return policy

Product Code. 10146990

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethylglyoxime, disodium salt octahydrate, 99%, C4H6N2Na2O2.8H2O, CAS Number-75006-64-3, c4h6n2o2.2na.8h2o, dimethylglyoxime di-sodium salt octahydrate, dimethylglyoxime disodium salt octahydrate, sodium dimethylglyoxime octahydrate, 1kg, White to Beige, 262-523-2, 304.19 | CAS: 75006-64-3 | C4H22N2Na2O10 | 304.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75006-64-3
Molecular Formula C4H22N2Na2O10
Molecular Weight (g/mol) 304.20
MDL Number MFCD00150288
InChI Key IJJDRGUPRATWQX-UHFFFAOYSA-L
Synonym dimethylglyoxime disodium salt octahydrate, c4h6n2o2.2na.8h2o, sodium dimethylglyoxime octahydrate, dimethylglyoxime di-sodium salt octahydrate
PubChem CID 71310106
SMILES O.O.O.O.O.O.O.O.[Na+].[Na+].CC(=N[O-])C(C)=N[O-]

Specifications

Melting Point >300.0°C
Color White to Beige
Loss on Drying 46% to 49%
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C(NONa)C(NONa)CH3·8H2O
Quantity 250 g
Solubility Information (50mg/ml,50% ethanol) Clear solution
IUPAC Name disodium;2-N,3-N-dioxidobutane-2,3-diimine;octahydrate
Formula Weight 304.19
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material Dimethylglyoxime, disodium salt octahydrate, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.