Learn More
Dimethylphenylphosphine, 96%, Thermo Scientific Chemicals
£99.70 - £355.00
Chemical Identifiers
CAS | 672-66-2 |
---|---|
Molecular Formula | C8H11P |
Molecular Weight (g/mol) | 138.15 |
MDL Number | MFCD00008509 |
InChI Key | HASCQPSFPAKVEK-UHFFFAOYSA-N |
Synonym | dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine |
PubChem CID | 69597 |
ChEBI | CHEBI:30671 |
IUPAC Name | dimethyl(phenyl)phosphane |
SMILES | CP(C)C1=CC=CC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
672-66-2 | |
138.15 | |
HASCQPSFPAKVEK-UHFFFAOYSA-N | |
69597 | |
dimethyl(phenyl)phosphane |
C8H11P | |
MFCD00008509 | |
dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine | |
CHEBI:30671 | |
CP(C)C1=CC=CC=C1 |
Specifications
672-66-2 | |
0.9710g/mL | |
50°C | |
97% | |
C6H5P(CH3)2 | |
MFCD00008509 | |
dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine | |
CP(C)C1=CC=CC=C1 | |
138.15 | |
CHEBI:30671 | |
97% | |
Dimethylphenylphosphine |
Colorless | |
74.0°C to 75.0°C (12.0 mmHg) | |
Authentic | |
C8H11P | |
1.5620 to 1.5640 | |
0.971 | |
HASCQPSFPAKVEK-UHFFFAOYSA-N | |
dimethyl(phenyl)phosphane | |
69597 | |
138.15 | |
Liquid |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 211-595-3
RUO – Research Use Only