missing translation for 'onlineSavingsMsg'
Learn More

Dimethylphenylphosphine, 96%

Product Code. 10519843 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10519843

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 672-66-2 | C8H11P | 138.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 672-66-2
Molecular Formula C8H11P
Molecular Weight (g/mol) 138.15
MDL Number MFCD00008509
InChI Key HASCQPSFPAKVEK-UHFFFAOYSA-N
Synonym dimethylphenylphosphine, dimethyl phenyl phosphine, dimethyl phenyl phosphane, me2pph, phenyldimethylphosphine, pme2ph, phosphine, dimethylphenyl, pubchem6420, dimethyl phenylphosphine
PubChem CID 69597
ChEBI CHEBI:30671
IUPAC Name dimethyl(phenyl)phosphane
SMILES CP(C)C1=CC=CC=C1

Specifications

Color Colorless
Density 0.9710g/mL
Boiling Point 74.0°C to 75.0°C (12.0 mmHg)
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C6H5P(CH3)2
Refractive Index 1.5620 to 1.5640
Quantity 25 mL
Specific Gravity 0.971
Formula Weight 138.15
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Dimethylphenylphosphine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.