missing translation for 'onlineSavingsMsg'
Learn More
Dimethylphenylsilane, 97%, Thermo Scientific Chemicals Product Code.: 11301627

Dimethylphenylsilane, 97%, Thermo Scientific Chemicals

Product Code. 11301627
50 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11301627

Brand: Thermo Scientific Alfa Aesar L04558.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Dimethylphenylsilane is a reagent for enol ether synthesis. It acts as a catalyst. Also used as a precursor in Optical Emission Spectroscopy of Plasma Deposition Processes. It can react with vinylbenzene to produce (b-Phenyl-ethyl)-dimethylphenyl-silan.

Solubility
Not miscible in water.

Notes
Moisture sensitive. Incompatible materials are Strong oxidizing agents, Strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 766-77-8
Molecular Formula C8H11Si
Molecular Weight (g/mol) 135.261
MDL Number MFCD00008257
InChI Key OIKHZBFJHONJJB-UHFFFAOYSA-N
Synonym dimethylphenylsilane, dimethyl phenyl silane, phenyldimethylsilane, silane, dimethylphenyl, benzene, dimethylsilyl, dimethylphenyl silane, c8h12si, dimethyl phenyl silyl, dimethyl-phenylsiliconShow MoreShow Less
PubChem CID 6327656
IUPAC Name dimethyl(phenyl)silicon
SMILES C[Si](C)C1=CC=CC=C1

Specifications

Boiling Point 156°C to 157°C
Melting Point -124°C
Quantity 50 g
UN Number UN1993
Beilstein 2204906
Flash Point 35°C (95°F)
Solubility Information Not miscible in water.
Formula Weight 136.27
Percent Purity 97%
Refractive Index 1.497
Density 0.879
Chemical Name or Material Dimethylphenylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberS25G033 Certificate TypeCertificate of Analysis Date04/11/2022 Catalog NumberL04558.09,L04558.18
Lot Number10173489 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL04558.09,L04558.18
Lot NumberR17C029 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04558.09,L04558.18
Lot Number10182129 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL04558.09,L04558.18
Lot NumberJ08T053 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL04558.09,L04558.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethylphenylsilane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.