missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsilyldiethylamine, 95%, Thermo Scientific Chemicals

£48.30 - £156.00

Chemical Identifiers

CAS 13686-66-3
Molecular Formula C6H16NSi
Molecular Weight (g/mol) 130.29
MDL Number MFCD00048523
InChI Key ADTGAVILDBXARD-UHFFFAOYSA-N
Synonym dimethylsilyldiethylamine, n,n-diethyl-1,1-dimethylsilylamine, diethylamino dimethylsilyl, silanamine, n,n-diethyl-1,1-dimethyl, silanamine,n,n-diethyl-1,1-dimethyl, diethylamino dimethyl silicon, diethylaminodimethylsilane, n,n-diethyl-1,1-dimethylsilanamine
PubChem CID 6335300
IUPAC Name (diethylamino)dimethylsilyl
SMILES CCN(CC)[Si](C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332186
View Documents
Thermo Scientific Alfa Aesar
L03503.06
5 g
£48.30
5g
Please to purchase this item. Need a web account? Register with us today!
11342186
View Documents
Thermo Scientific Alfa Aesar
L03503.14
25 g
£156.00
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13686-66-3
130.29
ADTGAVILDBXARD-UHFFFAOYSA-N
6335300
CCN(CC)[Si](C)C
C6H16NSi
MFCD00048523
dimethylsilyldiethylamine, n,n-diethyl-1,1-dimethylsilylamine, diethylamino dimethylsilyl, silanamine, n,n-diethyl-1,1-dimethyl, silanamine,n,n-diethyl-1,1-dimethyl, diethylamino dimethyl silicon, diethylaminodimethylsilane, n,n-diethyl-1,1-dimethylsilanamine
(diethylamino)dimethylsilyl

Specifications

109°C to 110°C
C6H16NSi
UN1993
<−5°C (23°F)
ADTGAVILDBXARD-UHFFFAOYSA-N
(diethylamino)dimethylsilyl
6335300
95%
Moisture sensitive
Dimethylsilyldiethylamine
13686-66-3
MFCD00048523
741862
dimethylsilyldiethylamine, n,n-diethyl-1,1-dimethylsilylamine, diethylamino dimethylsilyl, silanamine, n,n-diethyl-1,1-dimethyl, silanamine,n,n-diethyl-1,1-dimethyl, diethylamino dimethyl silicon, diethylaminodimethylsilane, n,n-diethyl-1,1-dimethylsilanamine
CCN(CC)[Si](C)C
130.29
131.3
1.408
0.8
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H225-H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

missing translation for 'einecsNumber' : 237-202-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only