missing translation for 'onlineSavingsMsg'
Learn More

Dimethylsuberimidate dihydrochloride

Product Code. 11329338
Click to view available options
Quantity:
2 g
10 g
Unit Size:
10g
2g
This item is not returnable. View return policy

Product Code. 11329338

Brand: Thermo Scientific Alfa Aesar 043244.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslinking of proteins and poly(allylamine) | CAS: 34490-86-3 | C10H22Cl2N2O2 | 273.198 g/mol

Dimethylsuberimidate dihydrochloride is used as a homobifunctional cross-linking reagent for amino groups. It is also used for the modification of tyrosinase to produce biocatalytic enzymatic flow-through detectors. It serves as a cross-linker for preparation of collagen-calcium phosphate composites. Further, it is used in the treatment of avian myeloblastosis virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 34490-86-3
Molecular Formula C10H22Cl2N2O2
Molecular Weight (g/mol) 273.198
MDL Number MFCD00012574
InChI Key ILKCDNKCNSNFMP-UHFFFAOYSA-N
Synonym dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochloride
PubChem CID 118696
IUPAC Name dimethyl octanediimidate;dihydrochloride
SMILES COC(=N)CCCCCCC(=N)OC.Cl.Cl

Specifica

Melting Point 213°C to 214°C
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 273.2
Physical Form Crystalline
Chemical Name or Material Dimethylsuberimidate dihydrochloride

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato