missing translation for 'onlineSavingsMsg'
Learn More
Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals Product Code.: 11339338

Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals

Product Code. 11339338
2 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 11339338

Brand: Thermo Scientific Alfa Aesar 043244.04

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Crosslinking of proteins and poly(allylamine)

Dimethylsuberimidate dihydrochloride is used as a homobifunctional cross-linking reagent for amino groups. It is also used for the modification of tyrosinase to produce biocatalytic enzymatic flow-through detectors. It serves as a cross-linker for preparation of collagen-calcium phosphate composites. Further, it is used in the treatment of avian myeloblastosis virus.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34490-86-3
Molecular Formula C10H22Cl2N2O2
Molecular Weight (g/mol) 273.198
MDL Number MFCD00012574
InChI Key ILKCDNKCNSNFMP-UHFFFAOYSA-N
Synonym dimethyl suberimidate dihydrochloride, octanediimidic acid, dimethyl ester, dihydrochloride, dimethyl octanebis imidate dihydrochloride, suberimidic acid dimethyl ester dihydrochloride, octanediimidic acid, 1,8-dimethyl ester, hydrochloride 1:2, 1,8-dimethyl octanediimidate dihydrochloride, suberimidic acid, dimethyl ester, dihydrochloride, acmc-1aiyg, 1,8-dimethoxyoctane-1,8-diyldiammonium dichloride, dimethylsuberimidate dihydrochlorideShow MoreShow Less
PubChem CID 118696
IUPAC Name dimethyl octanediimidate;dihydrochloride
SMILES COC(=N)CCCCCCC(=N)OC.Cl.Cl

Specifications

Melting Point 213°C to 214°C
Quantity 2 g
Sensitivity Moisture sensitive
Solubility Information Soluble in water.
Formula Weight 273.2
Physical Form Crystalline
Chemical Name or Material Dimethylsuberimidate dihydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM03L008 Certificate TypeCertificate of Analysis Date06/01/2025 Catalog Number043244.09,043244.04
Lot NumberM18I028 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043244.09,043244.04
Lot NumberX14A003 Certificate TypeCertificate of Analysis Date28/12/2021 Catalog Number043244.09,043244.04
Lot NumberW09A008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number043244.09,043244.04
Lot NumberG27K04 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number043244.09,043244.04

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethylsuberimidate dihydrochloride, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.