missing translation for 'onlineSavingsMsg'
Learn More
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals Product Code.: 10113161

Dimethyltin dichloride, 95%, Thermo Scientific Chemicals

Product Code. 10113161
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 10113161

Brand: Thermo Scientific Acros 347820050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 753-73-1
MDL Number MFCD00000517
InChI Key PKKGKUDPKRTKLJ-UHFFFAOYSA-L
Synonym dimethyltin dichloride, dimethyldichlorotin, stannane, dichlorodimethyl, dichlorodimethyltin, dimethyltindichloride, dimethyldichlorostannane, dichloro dimethyl stannane, tin, dimethyl-, dichloride, cotin 210, dichlorid dimethylcinicityShow MoreShow Less
PubChem CID 12955
IUPAC Name dichloro(dimethyl)stannane
SMILES C[Sn](C)(Cl)Cl

Specifications

Melting Point 103.0°C to 110.0°C
Color White to Yellow
Boiling Point 188.0°C to 190.0°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (Argentometry)
Packaging Glass bottle
Molecular Formula C2H6Cl2Sn
Linear Formula (CH3)2Sn(Cl)2
Quantity 5 g
Solubility Information Solubility in water: 20g/L (20°C). Other solubilities: 0.1 g/mL in methanol
Molecular Weight (g/mol) 219.66
Formula Weight 219.66
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material Dimethyltin dichloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0435824 Certificate TypeCertificate of Analysis Date10/11/2023 Catalog Number347820250,347821000,347820050
Lot NumberA0329781 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number347820250,347821000,347820050
Lot NumberA0388032 Certificate TypeCertificate of Analysis Date23/08/2021 Catalog Number347820250,347821000,347820050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Dimethyltin dichloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H372-Causes damage to organs.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
  • H361d-Suspected of damaging the unborn child.
  • H311-Toxic in contact with skin.
  • H301-Toxic if swallowed.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dimethyltin dichloride, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.