missing translation for 'onlineSavingsMsg'
Learn More

Dimethylzinc, 1.2M solution in toluene, AcroSeal™, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 377241000

132.00 GBP valid until 2024-07-31
Use promo code "23251" to get your promotional price.



Code : 29

Additional Details : CAS Number : 108-88-3 Weight : 0.09300kg Transport : UN number : 3399 Chem class : 4.3 Pack group : I


 View more versions of this product

Product Code. 10403073

  • £176.00 / 100mL
Estimated Shipment: 29-07-2024
to see stock.

This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Dimethylzinc
1.2M Solution in Toluene
C2H6Zn
MFCD00014854
0.9300g/mL
dimethylzinc, zinc, dimethyl, dimethyl zinc, methylzinc, unii-8h6r0n8w2f, ch3 2zn, znme2, zinc methyl, dimethylzink, hsdb 709
JRPGMCRJPQJYPE-UHFFFAOYSA-N
zinc;carbanide
11010
AcroSeal™ Glass bottle
100 mL
Clear colorless
544-97-8,108-88-3
Zn(CH3)2
1.2M (based on Zn) exact strength on the certificate of analysis
15, 3289
Solubility in water: vigorous reaction. Other solubilities: soluble in hydrocarbons
[CH3-].[CH3-].[Zn+2]
95.45
95.45
0.93
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Dimethylzinc, 1.2M solution in toluene

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Pyrophoric liquid Category 1
  • Substance which\, in contact with water\, emits flammable gases Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H304-May be fatal if swallowed and enters airways.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H361d-Suspected of damaging the unborn child.
  • H373-May cause damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH014-Reacts violently with water.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

Supplemental information
  • MIXTURE LIST-Contains : Toluene, Dimethyl zinc

SDS Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only