missing translation for 'onlineSavingsMsg'
Learn More

Dimidium bromide, 98%, Thermo Scientific Chemicals

£82.90 - £226.00

Chemical Identifiers

CAS 518-67-2
Molecular Formula C20H18BrN3
Molecular Weight (g/mol) 380.29
MDL Number MFCD00011757
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
Synonym dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
PubChem CID 68207
IUPAC Name 5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10174790
View Documents
Thermo Scientific Acros
215342500
250 mg Glass bottle
£82.90
250mg
Please to purchase this item. Need a web account? Register with us today!
10753724
View Documents Promotion Details
Thermo Scientific Acros
215340010
1 g Glass bottle
£226.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

518-67-2
380.29
MQOKYEROIFEEBH-UHFFFAOYSA-N
68207
[Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
C20H18BrN3
MFCD00011757
dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide

Specifications

518-67-2
Purple to Red
97.5% min. (on dry substance) (Argentometry)
MFCD00011757
09, 3257
Solubility in water: soluble
[Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
380.29
380.28
Powder
230°C
Authentic
C20H18BrN3
22, IV, 5515
dimidium bromide, trypadine, 3,8-diamino-5-methyl-6-phenylphenanthridinium bromide, phenanthridinium 1553, phenanthridinium compound 1553, phenanthridinium, 3,8-diamino-5-methyl-6-phenyl-, bromide, 2,7-diamino-10-methyl-9-phenylphenanthridinium bromide, 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide, phenanthridium 1553
MQOKYEROIFEEBH-UHFFFAOYSA-N
5-methyl-6-phenylphenanthridin-5-ium-3,8-diamine;bromide
68207
98%
Dimidium bromide, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 208-256-7

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only