missing translation for 'onlineSavingsMsg'
Learn More

Dimidium Bromide-Disulfine, Extra Pure, Blue Indicator Stock Solution, SLR, Fisher Chemical™

Product Code. 10283850 Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 mL
Packaging:
Amber glass bottle
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 10283850

Brand: Fisher Chemical D/4140/05

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Dimidium Bromide-Disulfine, Extra Pure, Blue Indicator Stock Solution, SLR, Fisher Chemical, Quantity: 100mL, Packaging: Amber glass bottle, Boiling Point: 100 deg.C, Melting Point: 0 deg.C, MDL Number: 146696, Physical Form: Liquid

TRUSTED_SUSTAINABILITY

Specifications

CAS 518-67-2
Melting Point 0°C
Color Blue
Boiling Point 100°C
Packaging Amber glass bottle
Molecular Formula C20H18BrN3
MDL Number 146696
Quantity 100 mL
InChI Key MQOKYEROIFEEBH-UHFFFAOYSA-N
SMILES [Br-].C[N+]1=C(C2=CC=CC=C2)C2=CC(N)=CC=C2C2=CC=C(N)C=C12
IUPAC Name 3,8-diamino-5-methyl-6-phenylphenanthridin-5-ium bromide
Molecular Weight (g/mol) 380.29
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.