Learn More
OHAUS™ Dimpled Mat
OHAUS™ Dimpled Mat designed to securely hold tubes in place.
Brand: OHAUS™ 30400143
Additional Details : Weight : 0.90000kg
Description
- A variety of platform options are available to hold irregular samples or traditional flask clamps and tube racks
- Platforms can increase the amount of usable space for samples
- In some cases can be stacked to increase throughput and double sample capacity
- Tube racks and dimpled mats securely hold tubes in place
Specifications
Platform Mat | |
9.25 in. | |
23.5 x 18.4 cm | |
23.50 cm | |
18.40 cm | |
Incubating Waver |
PVC | |
23.50 cm | |
9.25 in. | |
7.25 in. | |
Dimpled Mat, Incubating Waver ISWV02HDG |