Learn More
Lenz Laborglasintrumente Dimroth Condensers for Extractors, with Olives
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
£118.00 - £190.00
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Price | Quantity & Availability | |||||
10261981
|
Lenz Laborglasintrumente
05246008 |
71/51 NS |
£190.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
10393191
|
Lenz Laborglasintrumente
05246005 |
34/35 NS |
£118.00
Each |
Estimated Shipment: 15-08-2024
Log in to see stock. |
|||||
Description
- Dimroth condensers for extractors
- For condensing vapours
- With NS cone
- Water connection: glass olives
- Made of DURAN™ tubing
Specifications
Duran Glass | |
Glass olives | |
1 Ea. |