missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl sulfosuccinate, sodium salt, 96%

Product Code. 11935031 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
100g
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 11935031

Brand: Thermo Scientific Acros 117100025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-11-7 | C20H37NaO7S | 444.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
PubChem CID 23673837
IUPAC Name sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Acidity 2.5 max. (on solids basis)
Melting Point 153.0°C to 157.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min
Packaging Plastic drum
Quantity 2.5 kg
Beilstein 04, IV, 114
Fieser 15,149
Merck Index 15, 3446
Solubility Information Solubility in water: 1.5g/100mL (25°C). Other solubilities: soluble in ccl4,petroleum ether,xylene and,alcohol
Formula Weight 444.55
Percent Purity 96%
Insoluble Matter 300ppm (50% max. in toluene)
Physical Form Waxy Solid
Chemical Name or Material Dioctyl sulfosuccinate, sodium salt
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.