missing translation for 'onlineSavingsMsg'
Learn More

Dioctyl sulfosuccinate, sodium salt, 96%

Product Code. p-4509553 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic Bottle
Plastic drum
Unit Size:
100g
1kg
2.5kg
This item is not returnable. View return policy

Product Code. 10214480

Brand: Thermo Scientific Acros 117101000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 577-11-7 | C20H37NaO7S | 444.56 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 577-11-7
Molecular Formula C20H37NaO7S
Molecular Weight (g/mol) 444.56
MDL Number MFCD00012455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
Synonym docusate sodium, dioctyl sodium sulfosuccinate, aerosol ot, constonate, diox, manoxol ot, diomedicone, clestol, complemix, defilin
PubChem CID 23673837
IUPAC Name sodium;1,4-bis(2-ethylhexoxy)-1,4-dioxobutane-2-sulfonate
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O

Specifications

Acidity 2.5 max. (on solids basis)
Color White
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic Bottle
Quantity 100 g
Beilstein 04, IV, 114
Fieser 15,149
Merck Index 15, 3446
Solubility Information 300ppm max. Insoluble Matter (in toluene, in 50% soln.)
Formula Weight 444.55
Percent Purity ≥95%
Physical Form Waxy Solid
Chemical Name or Material Dioctyl sulfosuccinate, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • Dioctyl sulfosuccinate, sodium salt
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.