missing translation for 'onlineSavingsMsg'
Learn More
Dioctyl Sulfosuccinate Sodium Salt, Aerosol™ OT-100, Pure, Fisher Chemical Product Code.: 10010000

Dioctyl Sulfosuccinate Sodium Salt, Aerosol™ OT-100, Pure, Fisher Chemical

Product Code. 10010000
500g, Plastic powder jar
Click to view available options
500g, Plastic powder jar
This item is not returnable. View return policy

Product Code. 10010000

Brand: Fisher Chemical A/1040/53

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 153°C
CAS 577-11-7
Color White
Physical Form Solid
Quantity 500 g
Molecular Formula C20H37NaO7S
MDL Number 12455
InChI Key APSBXTVYXVQYAB-UHFFFAOYNA-M
SMILES [Na+].CCCCC(CC)COC(=O)CC(C(=O)OCC(CC)CCCC)S([O-])(=O)=O
IUPAC Name sodium 1,4-bis[(2-ethylhexyl)oxy]-1,4-dioxobutane-2-sulfonate
Molecular Weight (g/mol) 444.56
Formula Weight 444.55g/mol
Packaging Plastic powder jar
Show More Show Less
compliance-icons
Product Identifier
  • Dioctyl sulfosuccinate, sodium salt
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Dioctyl Sulfosuccinate Sodium Salt, Aerosol™ OT-100, Pure, Fisher Chemical >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.