Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Brand: Thermo Scientific™ 063419
Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
- Achieve n, n-1 resolution for oligonucleotides
- Resolve oligonucleotides with secondary structures
- Assay phosphorothioate purity
- Selectivity control with eluent pH, salt, and solvent
- Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications
Guard Column | |
HPAE-PAD | |
4000 psi (275 bar) | |
2.5 to 12.5 | |
Each | |
Quaternary Ammonium | |
Nonporous Polymer | |
Ion Exchange | |
Bio-LC |
9 mm | |
50 mm | |
8 μm | |
Nonporous | |
85°C | |
DNAPac | |
Ion Exchange | |
Guard Column |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.