Learn More
Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
Brand: Thermo Scientific™ 082508
Code : 15
Additional Details : Weight : 0.25000kg
Description
Enjoy strong anion exchange for high-resolution analysis and purification of synthetic oligonucleotides with Thermo Scientific™ DNAPac™ PA200 Oligonucleotide HPLC Columns.
- Achieve n, n-1 resolution for oligonucleotides
- Resolve oligonucleotides with secondary structures
- Assay phosphorothioate purity
- Selectivity control with eluent pH, salt, and solvent
- Resolve RNA with aberrant (2' – 5') links from normal SSRNA
Specifications
Analytical Column | |
UltiMate 3000 Titanium System | |
10000 psi (690 bar) | |
2.5 to 12.5 | |
85°C | |
DNAPac | |
Ion Exchange | |
Analytical Column |
4.6 mm | |
50 mm | |
4 μm | |
Nonporous | |
Quaternary Ammonium | |
Spherical, Nonporous Polymer | |
Ion Exchange | |
Bio-LC |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.